Sucrose (Brix) Standard 5% - 1.340264 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS05S
Supplier: REAGECON
Sucrose (Brix) Standard 5% - 1.340264 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS05S
Supplier: REAGECON
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
SSL Broth and OFS Agar
Supplier: DOEHLER GROUP
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Fuchsin solution, decolourised Reag. Ph. Eur. 1039401
Supplier: VWR Chemicals
Sodium salicylate ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals