ContactUsLinkComponent
Sucrose Standard 15% - 1.355679 (± 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pl ace. Extended shelf-life of 12 weeks 1 * 15 mL
Catalog # REGEBS15
Supplier:  REAGECON
Sucrose Standard 15% - 1.355679 (± 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pl ace. Extended shelf-life of 12 weeks 1 * 15 mL
Catalog # REGEBS15
Supplier:  REAGECON
Frequently Bought Together