1st Dissolution Fluid Dissolution Media Conentrate - 961.5ml of Conc. Dilutes 25L 1 * 6 L
Catalog # REGEDBCJP1DF-02
Supplier: REAGECON
1st Dissolution Fluid Dissolution Media Conentrate - 961.5ml of Conc. Dilutes 25L 1 * 6 L
Catalog # REGEDBCJP1DF-02
Supplier: REAGECON
Frequently Bought Together
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals