ContactUsLinkComponent
Hydrochloric Acid 0.01N - 961.5ml of Conc. Dilutes 25L 1 * 6 L
Catalog # REGEDBC10-960
Supplier:  REAGECON
Hydrochloric Acid 0.01N - 961.5ml of Conc. Dilutes 25L 1 * 6 L
Catalog # REGEDBC10-960
Supplier:  REAGECON
Frequently Bought Together