ORP BUFFER 271 MV, 500 ML 1 * 500 mL
HAMI238228
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
pH indicator papers, Duotest and Tritest
: MACHEREY-NAGEL
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Magnetic Stirrers, Boxed Sets
: VWR Collection
VWR®, Redox (ORP) Standards
: VWR Chemicals
Chemical scoops
: BOCHEM
pH Cleaning Solution Set
: HAMILTON BONADUZ