ContactUsLinkComponent
Laboratory chair neon, non slip, easy to clean, seat inclination adjustment, seat height adjustment with a gasspring from 450-620 mm , 5-star base made of aluminium, with glides with stop-and-go-castors, with foot ring. 1 * 1 items
Catalog # BIMOSP023279525807
Supplier:  bimos
Laboratory chair neon, non slip, easy to clean, seat inclination adjustment, seat height adjustment with a gasspring from 450-620 mm , 5-star base made of aluminium, with glides with stop-and-go-castors, with foot ring. 1 * 1 items
Catalog # BIMOSP023279525807
Supplier:  bimos
Frequently Bought Together