Order Entry
Austria
Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, Nitrite

VISOCOLOR® Colorimetric Test, Nitrite

VISOCOLOR® HE Nitrite, refill pack, Reagents for 150 tests in a box, 0,0 • 0,005 • 0,010 • 0,015 • 0,02 • 0,03 • 0,04 • 0,06 • 0,08 • 0,10 mg/L (NO₂⁻), Sulfanilic acid/1, Visual, 233×129×45 mm (including packaging), Hazardous material: Yes, 150, 1 KIT
Catalog # MANA920163
Supplier:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Nitrite
Catalog # MANA920163
Supplier:  MACHEREY-NAGEL
CAS Number:  
VISOCOLOR® Colorimetric Test, Nitrite
VISOCOLOR® Colorimetric Test, Nitrite

VISOCOLOR® HE Nitrite, refill pack, Reagents for 150 tests in a box, 0,0 • 0,005 • 0,010 • 0,015 • 0,02 • 0,03 • 0,04 • 0,06 • 0,08 • 0,10 mg/L (NO₂⁻), Sulfanilic acid/1, Visual, 233×129×45 mm (including packaging), Hazardous material: Yes, 150, 1 KIT

CAS Number:  
Price
76,80 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications

Specifications

  • Chemical method:
    Sulfanilic acid/1
  • Pk:
    1 KIT
  • Description:
    VISOCOLOR® HE Nitrite, refill pack
  • Dimensions:
    233×129×45 mm (including packaging)
  • Type:
    Hazardous material: Yes
  • For unit:
    Visual
  • Tests per kit:
    150
  • Range:
    0,0 • 0,005 • 0,010 • 0,015 • 0,02 • 0,03 • 0,04 • 0,06 • 0,08 • 0,10 mg/L (NO₂⁻)
  • Contents:
    Reagents for 150 tests in a box
  • Cat. No.:
    MANA920163
  • Platform:
    Colorimetric test kits
  • Model:
    MANA920163
  • Shelf life (months):
    24
  • Storage temperature:
    15…25 °C
  • Weight:
    240 g (including packaging)

Specifications