96/48 WELL COMBIBLOCK FOR GS-1 for 96x 0.2 ml tubes or 48x 0.5 ml tubes 1 * 1 ST
LBTHGS-B09648
: LABTECH
96/48 WELL COMBIBLOCK FOR GS-1 for 96x 0.2 ml tubes or 48x 0.5 ml tubes 1 * 1 ST
LBTHGS-B09648
: LABTECH
Frequently Bought Together
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Wasser, HiPerSolv CHROMANORM® für die HPLC
: VWR Chemicals
VWR® Gesichtstücher
: VWR Collection
VWR®, Nitrilhandschuhe
: VWR Collection
Nitrilhandschuhe, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Serologische Pipetten, Standard Serie
: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschuhe
: VWR Collection
VWR®, Transferpipetten, High Performance
: VWR Collection