Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Micro-fluidic platform, SilFlow™

Ferrules 0,35 mm ID, for use with tubing of OD 0,32 mm
  548-1800
Micro-fluidic platform, SilFlow™
  548-1800
Micro-fluidic platform, SilFlow™
Micro-fluidic platform, SilFlow™

Ferrules 0,35 mm ID, for use with tubing of OD 0,32 mm

 
230,00 €
   
1 * 10 ST
 

SilFlow™ microchannel device (MCD) options for three-port GC splitters, four-port GC splitters and a Deans' switch MCD, perfect for multi-dimensional analyses. SilFlow™ can be incorporated into the system without impacting the chromatography.

  • Chemically-deactivated stainless steel channels avoiding active sites experienced with conventional connections
  • Conventional graphite or Vespel® ferrules cannot be used with the SilFlow™ MCD as the dimensional stability is not adequate and there is a risk of contaminating the channels
  • SilTite™ FingerTite metal ferrules result in a reliable zero dead volume connection, giving optimised peak shapes
  • Can be operated at pressures greater than 25000 psi
  • SilFlow™ tracks oven temperature up to 20 °C/min. The design of SilFlow™ alleviates cold spots and sample condensation

 : SilFlow™ kits contain: SilFlow™ microchannel device (MCD), SilTite™ FingerTite tool, mounting bracket, appropriately sized ferrules, nuts and blanking ferrules for set up. 1,1 mm Ø tubing is not supplied in the splitter kits and must be ordered separately.

 
  • Description:
    Ferrules 0,35 mm ID, for use with tubing of OD 0,32 mm
  • Seq:
    16