Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Syringes for CTC analytics systems

Fixed needle, 26, 10 µl, 50 mm, 0,47 mm, Conical, 10F-BT-5/0.47C 6 pack
Catalog # 548-1938
Syringes for CTC analytics systems
Catalog # 548-1938
Syringes for CTC analytics systems
Syringes for CTC analytics systems

Fixed needle, 26, 10 µl, 50 mm, 0,47 mm, Conical, 10F-BT-5/0.47C 6 pack

Price
373,00 €
Log in for availability
1 * 6 ST
Specifications
  • Ext.Ø:
    0,47 mm
  • Gauge:
    26
  • Type:
    10F-BT-5/0.47C 6 pack
  • Type of tip:
    Conical
  • Volume:
    10 µl
  • Description:
    Fixed needle
  • Length:
    50 mm
  • Seq:
    30