Jump to:
Product Details & Documents
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:13,00837
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:190…192 °C
- Density:1.694 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ300
- Storage Temperature:Ambient
- EINECS:200-066-2
- REACH:exempt
Documents
Specifications
Specification Test Results
- Solubility:330 g/l (24 °C)
- Melting Point:190 - 192 °C (decomposition)
- Molar Mass:176.12 g/mol
- Bulk Density:500 - 900 kg/m3
- Vapor Pressure:(20 °C)
- Density:1.65 g/cm3 (20 °C)
- pH:2.2 - 2.5 (50 g/l, H2O, 20 °C)
- Ignition Point:380 °C
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



