Order Entry
Austria
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid

L(+)-Ascorbic acid

Plastic bottle, 100 g
Catalog # 1.00468.0100
Supplier:   Merck
L(+)-Ascorbic acid
Catalog # 1.00468.0100
Supplier:  Merck
CAS Number:  50-81-7
L(+)-Ascorbic acid
L(+)-Ascorbic acid

Plastic bottle, 100 g

CAS Number:  50-81-7
Price
102,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt

Documents

Specifications

Specification Test Results

  • Solubility:
    330 g/l (24 °C)
  • Melting Point:
    190 - 192 °C (decomposition)
  • Molar Mass:
    176.12 g/mol
  • Bulk Density:
    500 - 900 kg/m3
  • Vapor Pressure:
    (20 °C)
  • Density:
    1.65 g/cm3 (20 °C)
  • pH:
    2.2 - 2.5 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    380 °C

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.