Orders LinkContactUsLinkComponent
ClickSpeed mopping system
ClickSpeed mopping system
ClickSpeed mop frame, - mm, 400 mm
Catalog # VILE138621
Supplier:   FHP VILEDA SCS
ClickSpeed mopping system
Catalog # VILE138621
Supplier:  FHP VILEDA SCS
ClickSpeed mopping system
ClickSpeed mopping system
ClickSpeed mop frame, - mm, 400 mm
Price
39,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
ClickSpeed is the answer to customer requests for a floor cleaning system in the low to mid investment range. The ClickSpeed system has a unique and patented mop frame, which is very easy to handle and use. Simply drop the mop on the floor, slide in the frame and then click-close the magnetic mop holder. This means touch-less handling of dirty mops, which is beneficial to the user's hygiene.
  • Lightweight and simple to operate and use
  • Touchless mop attaching and removing
The nonwoven disposable mop contains microfibre and carries the patented novolon structure to pick up loose dirt. The mops are colour coded to avoid cross contamination. After use the mops are thrown away. ClickSpeed disposable mops cost as much as the wash cost for a durable mop in an external laundry.
The microfibre mop is a warp knitted high performance microfibre mop with polyester stripes. It is durable in use and washing. Working with it means saving force and time by achieving a better result.
Specifications
  • Description:
    ClickSpeed mop frame
  • Length:
    400 mm
  • L×W:
    - mm