Loop interfaces, 4 – 20 mA current
4 – 20 mA current loop interface, with temperature of 0 to +90 °C, UN30PA
Catalog # MEMMUN30PAV6
Supplier: MEMMERT
Springe zu:
Product Details & Documents
Spezifikationen
- Description:4 – 20 mA current loop interface, with temperature of 0 to +90 °C
- Seq:082
- For:UN30PA
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



