Loop interfaces, 4 – 20 mA current
4 – 20 mA current loop interface, with temperature of 0 to +90 °C, UN110PA
Catalog # MEMMUN110PAV6
Supplier: MEMMERT
Jump to:
Product Details & Documents
Specifications
- Description:4 – 20 mA current loop interface, with temperature of 0 to +90 °C
- Seq:088
- For:UN110PA
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



