Order Entry
Austria
Orders LinkContactUsLinkComponent
Loop interfaces, 4 – 20 mA current

Loop interfaces, 4 – 20 mA current

4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C, UN110PA
Catalog # MEMMUN110PAV3
Supplier:   MEMMERT
Loop interfaces, 4 – 20 mA current
Catalog # MEMMUN110PAV3
Supplier:  MEMMERT
Loop interfaces, 4 – 20 mA current
Loop interfaces, 4 – 20 mA current

4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C, UN110PA

Price
227,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C
  • Seq:
    078
  • For:
    UN110PA
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.