Order Entry
Austria
Orders LinkContactUsLinkComponent
Loop interfaces, 4 – 20 mA current

Loop interfaces, 4 – 20 mA current

4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C, UN75PA
Catalog # MEMMUN75PAV3
Supplier:   MEMMERT
Loop interfaces, 4 – 20 mA current
Catalog # MEMMUN75PAV3
Supplier:  MEMMERT
Loop interfaces, 4 – 20 mA current
Loop interfaces, 4 – 20 mA current

4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C, UN75PA

Price
227,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C
  • Seq:
    076
  • For:
    UN75PA