Orders LinkContactUsLinkComponent
VWR®, N-Plan Objectives
VWR®, N-Plan Objectives
IOS N-plan achromatic objective, 0,45 mm
Catalog # 630-1642
Supplier:   VWR Collection
VWR®, N-Plan Objectives
Catalog # 630-1642
Supplier:  VWR Collection
VWR®, N-Plan Objectives
VWR®, N-Plan Objectives
IOS N-plan achromatic objective, 0,45 mm
Price
102,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
Specifications
  • Magnification:
    40×/0,65 mm
  • Description:
    IOS N-plan achromatic objective
  • Seq:
    204
  • Working distance:
    0,45 mm