Order Entry
Austria
Orders LinkContactUsLinkComponent
J.T.Baker® BAKER ANALYZED®, Buffer Solutions

J.T.Baker® BAKER ANALYZED®, Buffer Solutions

10,00, ±0,05, Borate, 25 °C, Plastic bottle, 500 mL
Catalog # 5609.0500
Supplier:   Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
Catalog # 5609.0500
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
J.T.Baker® BAKER ANALYZED®, Buffer Solutions

10,00, ±0,05, Borate, 25 °C, Plastic bottle, 500 mL

CAS Number:  
Price
40,57 €
Einloggen für Verfügbarkeit
1 * 500 mL
Product Details & Documents

Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilise quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control.

Contain 0,01% Sodium benzoate.

Certifications: This buffer is traceable to NIST Standard reference buffer.

Spezifikationen

Spezifikationen

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0,05
  • pH value:
    10,00

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.