J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
10,00, ±0,05, Borat, Blau, 25 °C, 10 L
Supplier: Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
Catalog # 5658.9010
Supplier: Avantor
CAS Number:
Jump to:
Product Details & Documents
The quality of many BAKER ANALYZED™ reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS).
- Colour coded
- Potassium hydrogen phthalate based pH 4 buffer (red)
- Phosphate based pH 7 buffer (yellow)
- Borate based pH 10 buffer (blue)
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.
Solutions are available in both clear and colour-coded varieties.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Certifications: Each lot compared to NIST Standard Reference Buffer.
Documents
Certificates
Search by Lot NumberChemical Mining Supplies
Product Literature
spec_BAKR5647.0500.pdf spec_BAKR5647.1000.pdf spec_BAKR5647.9010.pdf spec_BAKR5655-01 spec_BAKR5656-03
Product specification
More
Specifications
Specifications
- pH-Puffertyp:Borat
- VE:10 L
- Referenztemperatur:25 °C
- Farbe:Blau
- pH Genauigkeit:±0,05
- pH-Wert:10,00



