Order Entry
Austria
Orders LinkContactUsLinkComponent
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded

J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded

4,00, ±0,02, Phthalate, Red, 20 °C, 500 mL
Catalog # 5676.0500
Supplier:   Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
Catalog # 5676.0500
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded

4,00, ±0,02, Phthalate, Red, 20 °C, 500 mL

CAS Number:  
Price
39,67 €
Log in for availability
1 * 500 mL
Product Details & Documents

The quality of many BAKER ANALYZED™ reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS).

  • Colour coded
  • Potassium hydrogen phthalate based pH 4 buffer (red)
  • Phosphate based pH 7 buffer (yellow)
  • Borate based pH 10 buffer (blue)

Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.

Solutions are available in both clear and colour-coded varieties.

A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Certifications: Each lot compared to NIST Standard Reference Buffer.

Specifications

Specifications

  • pH buffer type:
    Phthalate
  • Pk:
    500 mL
  • Reference temperature:
    20 °C
  • Colour:
    Red
  • pH accuracy:
    ±0,02
  • pH value:
    4,00

Specifications