Order Entry
Austria
Orders LinkContactUsLinkComponent
Forceps, precision

Forceps, precision

Straight, pointed, ultra fine tips, 0, 120 mm
Catalog # OUTI0
Supplier:   OUTILS RUBIS
Forceps, precision
Catalog # OUTI0
Supplier:  OUTILS RUBIS
Forceps, precision
Forceps, precision

Straight, pointed, ultra fine tips, 0, 120 mm

Price
27,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Forceps with very precisely machined tips. Ideal for applications that require a high degree of precision, including microscopy, anatomy, biology, microbiology, gene technology and electron microscopy.

  • Acid-resistant
  • Resistant against hydrogen fluoride and nitric acid
  • 100% anti-magnetic

Documents

Specifications
  • Type:
    0
  • Description:
    Straight, pointed, ultra fine tips
  • Length:
    120 mm