Order Entry
Austria
Orders LinkContactUsLinkComponent
Forceps, precision

Forceps, precision

Angled, pointed, fine tips, 6, 112 mm
Catalog # OUTI6
Supplier:   OUTILS RUBIS
Forceps, precision
Catalog # OUTI6
Supplier:  OUTILS RUBIS
Forceps, precision
Forceps, precision

Angled, pointed, fine tips, 6, 112 mm

Price
35,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Forceps with very precisely machined tips. Ideal for applications that require a high degree of precision, including microscopy, anatomy, biology, microbiology, gene technology and electron microscopy.

  • Acid-resistant
  • Resistant against hydrogen fluoride and nitric acid
  • 100% anti-magnetic

Documents

Specifications
  • Type:
    6
  • Description:
    Angled, pointed, fine tips
  • Length:
    112 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.