BUFFER SOLN REDOX UH = 427MV 220MV 25°C 1 * 250 ml
Catalog # METN51340065
Supplier: Mettler - Toledo
BUFFER SOLN REDOX UH = 427MV 220MV 25°C 1 * 250 ml
Catalog # METN51340065
Supplier: Mettler - Toledo
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
CHROMAFIL® Syringe Filters
Supplier: MACHEREY-NAGEL
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
BLAUBRAND® Volumetric Flasks, Class A, Blue Markings
Supplier: Brand
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Precision Grip Gloves with Dots
Supplier: VWR Collection
VWR®, Tubing, Silicone
Supplier: VWR Collection
Acetic acid 30% for biopharmaceutical production
Supplier: VWR Chemicals