Graduated pipettes
5 ml, 0,2 ml
Catalog # NEUB0251-28-220
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Glass, brown
Specifications
- Capacity:5 ml
- Seq:01
- Division:0,2 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



