Orders LinkContactUsLinkComponent
Isosorbide, Reference Standards
Isosorbide, Reference Standards
Diluted Isosorbide Mononitrate, USP reference standard, Vial, 750 mg
Catalog # 1353102.
Supplier:   USP
Isosorbide, Reference Standards
Catalog # 1353102.
Supplier:  USP
CAS Number:  
Isosorbide, Reference Standards
Diluted Isosorbide Mononitrate, USP reference standard, Vial, 750 mg
CAS Number:  
Price
466,00 €
Log in for availability
1 * 750 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Documents
Specifications
Specifications
  • Pack type:
    Vial
  • Pk:
    750 mg
  • Standards:
    USP reference standard
  • Description:
    Diluted Isosorbide Mononitrate
  • Cat. No.:
    1353102.
  • Description:
    Diluted Isosorbide Mononitrate
  • CAS No.:
    16051-77-7