Orders LinkContactUsLinkComponent
Polyoxyl stearate, Reference Standards
Polyoxyl stearate, Reference Standards
Polyoxyl 75 Stearate, USP reference standard, Vial, 200 mg
Catalog # 1547459.
Supplier:   USP
Polyoxyl stearate, Reference Standards
Catalog # 1547459.
Supplier:  USP
CAS Number:  
Polyoxyl stearate, Reference Standards
Polyoxyl 75 Stearate, USP reference standard, Vial, 200 mg
CAS Number:  
Price
236,00 €
Log in for availability
1 * 200 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications
Specifications
  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Polyoxyl 75 Stearate
  • Cat. No.:
    1547459.
  • Description:
    Polyoxyl 75 stearate
  • CAS No.:
    9004-99-3
Specifications