Order Entry
Austria
Orders LinkContactUsLinkComponent
Dantrolene, Reference Standards

Dantrolene, Reference Standards

Dantrolene Sodium, USP reference standard, Vial, 100 mg
Catalog # 1163151.
Supplier:   USP
Dantrolene, Reference Standards
Catalog # 1163151.
Supplier:  USP
CAS Number:  
Dantrolene, Reference Standards

Dantrolene Sodium, USP reference standard, Vial, 100 mg

CAS Number:  
Price
372,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    USP reference standard
  • Description:
    Dantrolene Sodium
  • Cat. No.:
    1163151.
  • Formula:
    C₂₈H₃₂N₈Na₂O₁₇
  • Description:
    Dantrolene Sodium
  • CAS No.:
    24868-20-0

Specifications