ContactUsLinkComponent
Complete ice-less cooling station to maintain samples at 0.5 to 4 °C for up to 18 hours without power. Includes CoolBox XT system, automation-compatible CoolRack XT PCR96 to hold one 96-well PCR plate or strip well tubes and one pack of 500 TruCool m 1 * 1 items
Catalog # 216-0968
Supplier:  BIOCISION
undefined
Complete ice-less cooling station to maintain samples at 0.5 to 4 °C for up to 18 hours without power. Includes CoolBox XT system, automation-compatible CoolRack XT PCR96 to hold one 96-well PCR plate or strip well tubes and one pack of 500 TruCool m 1 * 1 items
Catalog # 216-0968
Supplier:  BIOCISION
Frequently Bought Together