Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
100 ml, 19/26 NS
Catalog # LENZ04018037
Supplier:   Lenz Laborglas
Separating funnels, Squibb
Catalog # LENZ04018037
Supplier:  Lenz Laborglas
Separating funnels, Squibb
Separating funnels, Squibb
100 ml, 19/26 NS
Price
161,00 €
Log in for availability
1 * 2 ST
Product Details & Documents
Separating funnels, Squibb, stopcock with glass or PTFE key, made of borosilicate glass amberized.
  • Without graduation
  • Very good temperature and chemical resistance
  • Amber coloured for ligth sensitve liquids
Documents
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml