ContactUsLinkComponent
for effective sample disruption Compact and effective benchtop homogeniser for the disruption of a wide range of biological samples in 0.5, 2.0 or 7 ml tubes using bead beating technology. Extremely high lysis efficiency thanks to 3D motion. Fully co 1 * 1 items
Catalog # BERT06404.200RD_B
Supplier:  Bertin Instruments
for effective sample disruption Compact and effective benchtop homogeniser for the disruption of a wide range of biological samples in 0.5, 2.0 or 7 ml tubes using bead beating technology. Extremely high lysis efficiency thanks to 3D motion. Fully co 1 * 1 items
Catalog # BERT06404.200RD_B
Supplier:  Bertin Instruments
Frequently Bought Together