Orders LinkContactUsLinkComponent
Separating funnels, conical
Separating funnels, conical
100 ml, 19/26 NS
Catalog # 532-0104
Supplier:   Lenz Laborglas
Separating funnels, conical
Catalog # 532-0104
Supplier:  Lenz Laborglas
Separating funnels, conical
Separating funnels, conical
100 ml, 19/26 NS
Price
89,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Separating funnels, conical, made of borosilicate glass 3.3, amber coloured.
  • Amber coloured to protect light sensitive substances
  • Large labeling field
  • Very good chemical and temperature resistance
With interchangeable PTFE key.
Documents
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml