Measuring scoops
100 ml, Green
Catalog # 231-0414
Supplier: VITLAB
Springe zu:
Product Details & Documents
PP, coloured. With precision formed filling edge and comfortable, stable handle.
- Also suitable as weighing scoops
- Easily readable volume quantities visible on the upper side of the handle
- Food-safe product according to EC Directive No. 10/2011
Spezifikationen
- Capacity:100 ml
- Colour:Green