Measuring scoops
100 ml, Yellow
Artikelnr. 231-0412
Anbieter: VITLAB
Jump to:
Product Details & Documents
PP, coloured. With precision formed filling edge and comfortable, stable handle.
- Also suitable as weighing scoops
- Easily readable volume quantities visible on the upper side of the handle
- Food-safe product according to EC Directive No. 10/2011
Specifications
- Capacity:100 ml
- Colour:Yellow