ContactUsLinkComponent
Alaris Self venting 0.2µm(+ve) particle filter (with 22cm DEHP-free-PVC-extension tube), PES Supor HP-200(10cm2), positively charged - 1 x Pack of 50 1 * 50 items
Catalog # BDAMMFX1826
Supplier:  BD Medical
Alaris Self venting 0.2µm(+ve) particle filter (with 22cm DEHP-free-PVC-extension tube), PES Supor HP-200(10cm2), positively charged - 1 x Pack of 50 1 * 50 items
Catalog # BDAMMFX1826
Supplier:  BD Medical
Frequently Bought Together