Order Entry
Austria
Orders LinkContactUsLinkComponent
Bath fluids

Bath fluids

Descaling agent, based on citric acid, — °C, − °C, − °C, Colourless, 5 kg
Catalog # 461-1115
Supplier:   LAUDA
Bath fluids
Catalog # 461-1115
Supplier:  LAUDA
CAS Number:  
Bath fluids
Bath fluids

Descaling agent, based on citric acid, — °C, − °C, − °C, Colourless, 5 kg

CAS Number:  
Price
168,00 €
Log in for availability
1 * 5 kg
Product Details & Documents

Allow highly accurate thermostating, even at extreme temperatures.

  • Simple and safe handling, suitable for long term operation
  • Optimise thermostat life

* Denotes Silicone oil
** Only suitable for closed circuits

Specifications

Specifications

  • Pk:
    5 kg
  • Colour:
    Colourless
  • Fire point:
    − °C
  • Flash point:
    − °C
  • Temperature range:
    — °C
  • Type:
    Descaling agent, based on citric acid

Specifications