Product Details & Documents
Chromolith® Prep monolithic stationary phases are new ultrapure silica phases which gain their special properties from a bimodal pore structure. This structure is a property of the new 'Sol-Gel' technology and consists of macropores and mesopores. The mesopores with an average diameter of 12 nm, form the fine porous structure of the column interior and create a very large surface area on which adsorption of the target compounds occurs.
The large macropores, with a pore diameter ranging from 3 µm, form a dense network of pores and allow a high flow rate due to a low resistance factor. The resulting excellent accessibility of the mesopores (total porosity >80%) ensures fast adsorption and desorption kinetics because of the short diffusion length inside the pores. This results in dramatically reduced separation times and thus greatly increased productivity.
To operate a monolithic column it is necessary to ensure that no flow through occurs in the wall region. Therefore the monolith is cladded with a polymeric material (special PEEK) and can be connected directly to each HPLC system and used as 'ready to use' column.
Documents
Merck Chromolith® HPLC Columns
Product Literature
Specifications
- Column Format:Guard cartridge
- Ø int.:10,00 mm
- Description:Chromolith® SemiPrep RP-18 endcapped, guard column
- Length:10 mm
- Seq:06
- Phase:RP-18e
Accessories
Product Information
- Catalog #Ø int.LengthAvailabilityPrice
- Chromolith® SemiPrep guard cartridge holderChromolith® SemiPrep guard cartridge holder1.52037.000110,00 mm10 mmLog in for availability1 * 1 ST - 630,00 €



