ContactUsLinkComponent
5-Gas-Detektor as a Diffusion-Equipment Dust and Water Resistant: IP 65/67) in a protected Rubberboot Size: 5.7 x 2.9 x 1.5 in. / 14.5 x 7.4 x 3.8 cm Weight: 13.1 oz. / 370 g Temperature: -4 to +122°F / -20 to +50°C Sensors: O2: electrochemical Oxyge 1 * 1 items
Catalog # BWTEM5XWAYRDDYE00
Supplier:  BW TECHNOLOGIES
5-Gas-Detektor as a Diffusion-Equipment Dust and Water Resistant: IP 65/67) in a protected Rubberboot Size: 5.7 x 2.9 x 1.5 in. / 14.5 x 7.4 x 3.8 cm Weight: 13.1 oz. / 370 g Temperature: -4 to +122°F / -20 to +50°C Sensors: O2: electrochemical Oxyge 1 * 1 items
Catalog # BWTEM5XWAYRDDYE00
Supplier:  BW TECHNOLOGIES
Frequently Bought Together