ContactUsLinkComponent
14 COMPOSES Ca 150mg/l Mg 50mg/l Cd 10mg/l Na 100mg/l P 0.5mg/l B 0.5mg/l Zn 0.5mg/l Mn 0.2mg/l Fe 0.2mg/l Cr 0.2mg/l Al 0.2mg/l Cu 0.2mg/l Ni 0.2mg/l In 4mg/l DANS HNO3 1N 1 * 1.000 mL
Catalog # 89206.290
14 COMPOSES Ca 150mg/l Mg 50mg/l Cd 10mg/l Na 100mg/l P 0.5mg/l B 0.5mg/l Zn 0.5mg/l Mn 0.2mg/l Fe 0.2mg/l Cr 0.2mg/l Al 0.2mg/l Cu 0.2mg/l Ni 0.2mg/l In 4mg/l DANS HNO3 1N 1 * 1.000 mL
Catalog # 89206.290
Frequently Bought Together