Order Entry
Austria
Orders LinkContactUsLinkComponent
Wardrobe/filing cabinets | Avantor

Wardrobe/filing cabinets

800×420×2156 mm
Catalog # HAMBV268GA/W/W/WE
Supplier:   HAMMERBACHER
Wardrobe/filing cabinets
Catalog # HAMBV268GA/W/W/WE
Supplier:  HAMMERBACHER
Wardrobe/filing cabinets

800×420×2156 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Locker/combination cabinet with centre wall. Completely equipped with shelving on the left side, right clothes rail and two additional storage compartments. With clothes hook and door mirror. Carcass construction, i.e. each cabinet can stand on its own, carcass glued.

Material: Melamine resin coated chipboard in accordance with DIN 68765, emission class E1, all front edges covered with 2 mm ABS, rear edge of the carcass with 0,6 mm ABS. Sides and fronts 19 mm thick, base, panels and shelves 22 mm thick, rear panel 8 mm thick.
Height adjustment: With four plastic guides, 20 mm high, 80 mm diameter, adjustable from inside through the base of the cabinet (+10 mm).

Steel base (optional): 7 cm high, cabinet can be made 5,1 cm higher with the steel base than with the adjustable feet.

Doors: 2 swing doors with sealing lip, opening angle 125 degrees.

Panels/base: 22 mm thick, with 2 mm ABS edging on the front. Height adjustable in 32 mm increments with recessed shelf supports. Sides and fronts 19 mm thick.

Lock: 3-point locking mechanism with door damper buffer.
Hinges: High quality, all metal cup hinges manufactured by Hettich. Quick mounting technology, 3-dimensionally adjustable.

Delivery information: The cabinets are delivered fully assembled.

Specifications
  • Seq:
    14
  • W×D×H:
    800×420×2156 mm
Complementary Products

Product Information

  • Catalog #
    Description
    Availability
    Price
  • HAMBV2SO80/A
    Base, steel, for cabinets with 80 cm width
    Log in for availability
    1 * 1 ST - 0,00 €