Order Entry
Austria
ContactUsLinkComponent
DNA Polymerase Phusion Hot start II High-Fidelity (2U/µl) is supplied with 5x Phusion HF Buffer, 5x Phusion GC Buffer, DMSO and 50mM MgCl2 solution (100 reactions) 1 * 1 KIT
Phusion® Hot Start II high-fidelity DNA polymerase
Catalog # F-549S
Supplier:  FINNZYMES REAGENTS
CAS Number:  
undefined
Phusion® Hot Start II high-fidelity DNA polymerase
Catalog # F-549S
Supplier:  FINNZYMES REAGENTS
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Phusion® Hot Start II High-Fidelity DNA Polymerase, 100 U (2 U/µl)
  • Seq:
    01

Specifications

About this item

Phusion® Hot Start II High-Fidelity DNA Polymerase allows the use of a wide range of primers, including those with low melting temperatures. Phusion® Hot Start II High-Fidelity DNA Polymerase is ideal for PCR applications from routine PCR to highly demanding applications, such as cloning and high throughput PCR. It incorporates a unique dsDNA-binding domain making this Pyrococcus-like proofreading DNA polymerase extremely robust, rapid, and accurate.

  • No annealing temperature limitations
  • Allows reaction set up at room temperature
  • Zero-time reactivation due to unique Hot Start technology
  • Fast PCR enzyme with extreme fidelity (over 6 times more accurate than Pfu)
  • Robust enzyme with high tolerance against PCR inhibitors requires minimal optimisation
Frequently Bought Together