Kit PCR Phusion Human Specimen Direct 200x20µl reactions (contains Phusion Human Specimen DNA Polymerase, 2x Phusion Human Specimen PCR buffer (includes dNTPs and MgCl2), universal control primer mix (25µM each), dilution buffer, DNARelease additive) 1 * 1 KIT
Catalog # F-150
Supplier: FINNZYMES REAGENTS
Frequently Bought Together