ContactUsLinkComponent
System electroporation AgilePulse MAX large volume includes AgilePulse MAX pulse generator, safety stand, large volume electroporation stand, 6ml electroporation chamber (2 each), cables and manual 1 * 1 items
Catalog # 732-1503
Supplier:  BTX
System electroporation AgilePulse MAX large volume includes AgilePulse MAX pulse generator, safety stand, large volume electroporation stand, 6ml electroporation chamber (2 each), cables and manual 1 * 1 items
Catalog # 732-1503
Supplier:  BTX
Frequently Bought Together