BUFFER SOLUTION PH7.01 @25C 1 * 500 mL
Catalog # REGE70125050
Supplier: REAGECON
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions
Supplier: REAGECON
Supelco® Organic Reference Standard, 2,2,2-Trifluoroethanol
Supplier: Merck
PARACETAMOL IMPURITY J 1 * 15 mg
Supplier: EDQM
VWR®, Colour standards, Saybolt
Supplier: VWR Chemicals
Chloramphenicol, Supelco®
Supplier: Merck
3,3,5-Trimethylcyclohexyl salicylate certified 1 * 0,25 g
Supplier: EHRENSTORFER
VWR®, Turbidity standards, formazin
Supplier: VWR Chemicals
Buffers, concentrated solutions, Fluka™
Supplier: Honeywell Chemicals
Potassium permanganate 0.1 N in aqueous solution USP Test Solution (TS)
Supplier: VWR Chemicals