Order Entry
Austria
Orders LinkContactUsLinkComponent
Double spatulas, micro

Double spatulas, micro

150 mm, 40 mm, 2 mm
Catalog # WITG7.412.052
Supplier:   witeg Labortechnik
Double spatulas, micro
Catalog # WITG7.412.052
Supplier:  witeg Labortechnik
Double spatulas, micro
Double spatulas, micro

150 mm, 40 mm, 2 mm

Price
1,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Micro spatulas with two flat ends, made of stainless steel.

Specifications
  • Length:
    150 mm
  • Blade width:
    2 mm
  • Blade length:
    40 mm