Order Entry
Austria
Orders LinkContactUsLinkComponent
Temperature probes

Temperature probes

Needle probe, Type 80106, 0…600 °C, 3×130 mm
Catalog # SIMETE5106
Supplier:   Industrial Physics
Temperature probes
Catalog # SIMETE5106
Supplier:  Industrial Physics
Temperature probes
Temperature probes

Needle probe, Type 80106, 0…600 °C, 3×130 mm

Price
148,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These temperature probes can be connected to instruments suited for changeable probes (type ‘K’) using a plug connection.

  • Provided with a firm hand-grip and a one meter curled cable
  • Thicker probes have a longer response time

Documents

Specifications
  • Range:
    0…600 °C
  • Description:
    Needle probe
  • Seq:
    04
  • Type:
    Type 80106
  • Ø×L:
    3×130 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.