Test charts
Test charts with optical brightner, White/black, A6, 105×148 mm
SIMEVF2343
Test charts are used for testing physical properties of coating, lacquers, and inks.
- Dimensions from A6 up to A4
- Film laminated for an excellent solvent, chemical resistance, and film spread
- Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Ordering information: A certificate is available for charts with an optical brightener.
Documents
- Format:A6
- Colour:White/black
- Description:Test charts with optical brightner
- Seq:02
- W×H:105×148 mm