Orders LinkContactUsLinkComponent
Test charts
Test charts
Test charts with optical brightner, Black/white chequered, A4, 210×297 mm
Catalog # SIMEVF2347
Supplier:   Industrial Physics
Test charts
Catalog # SIMEVF2347
Supplier:  Industrial Physics
Test charts
Test charts
Test charts with optical brightner, Black/white chequered, A4, 210×297 mm
Price
184,00 €
Log in for availability
1 * 250 ST
Product Details & Documents
Test charts are used for testing physical properties of coating, lacquers, and inks.
  • Dimensions from A6 up to A4
  • Film laminated for an excellent solvent, chemical resistance, and film spread
  • Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Ordering information: A certificate is available for charts with an optical brightener.
Specifications
  • Format:
    A4
  • Colour:
    Black/white chequered
  • Description:
    Test charts with optical brightner
  • Seq:
    10
  • W×H:
    210×297 mm