Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Grindometers

Grindometer, double groove, 0 - 100 µm, graduation 10 µm
  SIMEVF2113
Grindometers
  SIMEVF2113
Grindometers
Grindometers

Grindometer, double groove, 0 - 100 µm, graduation 10 µm

 
536,00 €
   
1 * 1 ST
 

These grindometers or gauges of fineness are precision instruments to determine particle size and fineness of many materials like paints, lacquers, pigments, filler, chocolate and more. TQC Grindometers have double grooves with graded slopes graduated in three different parameters: µm (microns), NS (Hegman) and PCU (North).

  • Gauge and beveled scraper are made of hardened stainless steel and have an accuracy of 2 µm

W×D×H: 175×60×12 mm

 : Conforms to DIN EN ISO NF21524.

 : Supplied with test certificate.

 
  • Description:
    Grindometer, double groove, 0 - 100 µm, graduation 10 µm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.