Orders LinkContactUsLinkComponent
Specific gravity cups, calibrated
Specific gravity cups, calibrated
Anodised aluminium, 100 ml
Catalog # SIMEVF2097
Supplier:   Industrial Physics
Specific gravity cups, calibrated
Catalog # SIMEVF2097
Supplier:  Industrial Physics
Specific gravity cups, calibrated
Specific gravity cups, calibrated
Anodised aluminium, 100 ml
Price
321,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specific gravity cups for determining the specific gravity or density (or weight per gallon wpg) of coatings, pastes or similar liquids.
  • Density is defined as weight per unit volume at a specified temperature
  • Available in anodised aluminium or stainless steel
Certifications: According to ISO 2811, DIN 53 217 and ASTM D 1475.
Delivery information: Supplied with a calibration certificate.
Documents
Specifications
  • Volume (nom.):
    100 ml
  • Description:
    Anodised aluminium
  • Seq:
    02